man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Chromatography Columns and Cartridges
Ion Chromatography Columns
Anion-Exchange Packed IC Columns
Hamilton™ PEEK RCX-30
10186870
Print
Hamilton™ PEEK RCX-30
Column capacity: 1.2meq/column
Brand:
Hamilton™ 79370
View more versions of this product
Product Code.
10186870
1205.00 €
/ Each
Quantity
Estimated Shipment: 24-10-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Specifications
Column Capacity
1.2 meq/column
Particle Size
7 μm
Stationary Phase
RCX-30