Learn More
Thermo Scientific™ PEEK Union Adapter Assembly
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Brand:  Thermo Scientific™ P-742-01
Product Code. 15967881
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Absolute zero or low dead volume formats
  • Biocompatible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
10-32
For 1/16 in. Tubing O.D.
5000 psi
(2) F-300 Fingertight Fittings
Union Adapter Assembly
PEEK
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ PEEK Union Adapter Assembly > PEEK™ Unions BODY