missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Pehanon™ pH Paper

Measure pH in colored solutions. Pehanon™ pH paper puts the indicator and reference on one strip making it perfect for pH determinations in colored solutions.

17.40 € - 21.05 €

Specifications

Format Stick
For Use With (Application) Determination of pH in the colored solutions
Type pH Paper
View More Specs

Products 11
Product Code Brand pH Range Graduations Price Quantity & Availability  
Product Code Brand pH Range Graduations Price Quantity & Availability  
11366464
View Documents
Macherey-Nagel™
90415
3.8 to 5.5 3.8, 4.0, 4.2, 4.4, 4.6, 4.9, 5.2, 5.5
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10004760
View Documents
Macherey-Nagel™
90422
10.5 to 13 10.5, 11.0, 11.5, 12.0, 12.5, 13.0
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10196160
View Documents
Macherey-Nagel™
90416
5.2 to 6.8 5.2, 5.5, 5.7, 5.9, 6.1, 6.3, 6.5, 6.8
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10547502
View Documents
Macherey-Nagel™
90424
4 to 9 4.0, 4.5, 5.0, 5.5, 6.0, 6.5, 7.0, 7.5, 8.0, 8.5, 9.0
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10708911
View Documents
Macherey-Nagel™
90417
6 to 8.1 6.0, 6.3, 6.6, 6.9, 7.2, 7.5, 7.8, 8.1
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10058100
View Documents
Macherey-Nagel™
90414
2.8 to 4.6 2.8, 3.1, 3.4, 3.6, 3.8, 4.0, 4.3, 4.6
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10736991
View Documents
Macherey-Nagel™
90420
8 to 9.7 8.0, 8.2, 8.4, 8.6, 8.8, 9.1, 9.4, 9.7
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
11817043
View Documents
Macherey-Nagel™
90401
1 to 12 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12
21.05 €
Pack of 200
In Stock
to see stock.
Edge
Add to basket
 
10460431
missing translation for 'viewDocuments'
Macherey-Nagel™
90412
1 to 2.8 1.0, 1.3, 1.6, 1.8, 2.0, 2.2, 2.5, 2.8
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10741601
missing translation for 'viewDocuments'
Macherey-Nagel™
90419
7.2 to 8.8 7.2, 7.4, 7.6, 7.8, 8.0, 8.2, 8.5, 8.8
18.45 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
10687901
missing translation for 'viewDocuments'
Macherey-Nagel™
90421
9.5 to 12 9.5, 10.0, 10.5, 11.0, 11.5, 12.0
17.40 €
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Measure pH in colored solutions. Pehanon™ pH paper puts the indicator and reference on one strip making it perfect for pH determinations in colored solutions.

PEHANON™ is a special pH-test strips that unifies the pH-indicator and the reference colors on one strip and has the following advantages:

Any sample color has the same effect on both, the reference colors and the reactive pad. This ensures unadulterated readings in colored solutions. The user can be sure to get accurate pH-values.

An invisible hydrophobic barrier just above the top color field prevents capillary action of the test solution beyond. The handle will remain dry and clean whatever the sample is. Therefore the use of the strip is very safe.

pH-values can be determined without a separate color chart. Workers and machinists can use single strips instead of complete packs with color chart which makes PEHANON™ highly economic.
Spezifikation

Spezifikation

Stick
Determination of pH in the colored solutions
pH Paper
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt