missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12053-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody may cross react with Pellino-3.
The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.Specifications
PELI1 | |
Polyclonal | |
Unconjugated | |
PELI1 | |
PELI1, Pellino 1, pellino homolog 1 (Drosophila), PRISM, Protein pellino homolog 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305549, 57162, 67245 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C669, Q96FA3 | |
PELI1 | |
PELI1 Fusion Protein Ag2683 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |