Learn More
3M™ PELTOR™ Earmuffs
PELTOR™ X2 Earmuffs are our innovative hearing protectors with a modern slim cup design.
352.00 € - 363.00 €
Specifications
Noise Reduction Rating (NRR) 24 dB
Corded Uncorded
Plug Shape Oval
Disposable Reusable
Metal Detectable No
View More Specs
Products 2
Product Code Brand Style Price Quantity & Availability  
Product Code Brand Style Price Quantity & Availability  
17863276
View Documents
3M™
7000103990
Hard Hat
352.00 €
Pack of 10
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
17893246
View Documents
3M™
7100123157
Neckband
363.00 €
Pack of 10
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
Description
Description
  • For high performance hearing protection against moderate noise levels, use 3M™ PELTOR™ X2 Earmuffs.
  • 3M PELTOR Earmuffs X2 are colour-coded yellow, making it simple to select the correct level of protection from the X series range.
  • The headband model features an electrically insulated, twin headband design that helps reduce heat build-up on top of the head, and the stainless steel wire, coated with plastic makes the headband the most robust 3M has to offer.
  • It is both strong and comfortable as the flat wide headband has a soft plastic coating providing stable pressure on top of the head.
  • Carefully moulded sliders ensure the earmuffs are easy to adjust when on the head.
  • Wide, soft foam ear cushions help workers stay comfortable, while the slim cups offer improved compatibility with other safety equipment.
  • Offering a product to meet each users job or preference the helmet mounted version is available as P5 with the same dielectric performance as the headband model.
  • Alternatively, if the application does not require electrical insulation the helmet mounted P3 model and neckband is available.
  • The cushions and foam inserts, available as Hygiene Kits, are easy to replace if they become worn or damaged, helping extend the life of the product.
Specifications
Specifications
24 dB
Oval
No
No
Uncorded
Reusable
No