Learn More
3M™ PELTOR™ Optime™ I Earmuffs
PELTOR™ Optime™ I Ear Muffs are very lightweight, providing high wearer comfort. Ideal for protection against noise arising from a wide range of applications in the workplace and leisure activities. Offer versatile protection for both short and long duration tasks.
Brand:  3M™ 7000038206
205.56 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15709697
  • 286.00 € / Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low-profile design which minimizes obtrusiveness
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts helps keep them hygienically clean
  • Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hi-Viz
H510P3E-469-GB
Ear Muffs
CE Approved, EN 352-3: 2002
26 dB
Hard Hat