Learn More
3M™ PELTOR™ Optime™ II Ear Muffs Product Code.: 11729315
Add to basket
3M™ PELTOR™ Optime™ II Ear Muffs
Product Code. 11729315
Passive Headset
Click to view available options
Style:
Headband
Passive Headset
Product Code. 11729315
Brand: 3M™ 7000039619
Add to basket
Add to basket
Developed for demanding, noisy environments and particularly suited for noise levels dominated by low to medium frequencies. 3M™ PELTOR™ Optime™ II Ear Muffs provide ideal hearing protection in environments with substantial industrial noise or construction machinery, like airports and agricultural work.
  • Electrically insulated headband (dielectric) version available
  • Large space inside cup helps reduce moisture and heat build-up
  • Sealing rings filled with a unique combination of liquid and foam
  • Easy-to-replace cushions and inserts helps keep them hygienically clean
  • Soft, wide cusions help reduce pressure around the ears and improves comfort and wearability
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Certifications/Compliance CE Approved
For Use With (Application) Suitable Choice for Noisy Industrial Environments for the Agricultural Sector, Airports or for the Use of Construction Equipment
Plug Shape Oval
Product Type Hearing Protection Optime 2
Material Stainless Steel
Style Passive Headset
Product Content Correction
Product Title
3M™ PELTOR™ Optime™ II Ear Muffs >