Learn More
3M™ PELTOR™ Optime™ II Helmet Mounted Ear Muffs
Product Code. 12980295
Click to view available options
Color:
Black
Specifications
Color Black
Includes P3E Helmet Attachment for Helmet Mounting
Certifications/Compliance CE Approved
For Use With (Application) Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work
Noise Reduction Rating (NRR) 30 dB
Plug Shape Oval
Product Type Ear Muffs Optime II
Dielectric Yes
Material Stainless Steel
Style Helmet-mounted