missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ Optime™ II Helmet Mounted Ear Muffs
39.90 € - 730.00 €
Specifications
Color | Black |
---|---|
For Use With (Application) | Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work |
Plug Shape | Oval |
Style | Helmet-mounted |
Description
Specifications
Black | |
Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work | |
Oval | |
Helmet-mounted |