missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ Power Supply
3M™ PELTOR™ Power Supply is suitable for use with 3M™ PELTOR™ ACK053 rechargeable battery pack.
Brand: 3M™ 7000108521
This item is not returnable.
View return policy
Description
- Mains power socket plug with USB port
- Power adaptor 230 V DC to USB 5 V
- Compatible USB Charging Cable 3M™ PELTOR™ AL2AI or PELTOR™ FR09 required
- Supplied with EU/UK plug adapters
Specifications
Power Supply | |
Lite-Com™ Pro II IS Battery | |
USB Charging Port |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction