missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ X5 Series Ear Muffs
Click to view available options
Model:
X5A
Dielectric:
Yes
Style:
Over-the-head
Description
- Unparalleled attenuation without the need for double protection
- Offered in over-the-head or hard hat-attached styles
- Twin headband design helps reduce heat buildup with good fit and balance
- Combination of new technologies allows for more room inside the ear cup for a more comfortable fit
- Ear cup pivot points tilt for optimum comfort and efficiency
- Dielectric (models X5A and X5P5E only)
- Compatibility with hard hats (models X5P3E and X5P5E)
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications
Specifications
Color | Black |
ANSI S3.1974 | Yes |
Cushion Material | Foam |
CSA Z94.2 | Class AL |
Cushion Color | Black |
Headband Material | Wire |
For Use With (Application) | Composite Finishing, Grinding, Machine Operations, Paper Mills, Quarrying, Transportation, Welding |
Model | X5A |
Pairs per Package | 1 Ea. |
Noise Reduction Rating (NRR) | 31 dB |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ PELTOR™ X5 Series Ear Muffs >
Spot an opportunity for improvement?Share a Content Correction