missing translation for 'onlineSavingsMsg'
Learn More

Pentacene, 97%

Product Code. 15402778
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15402778

Brand: Thermo Scientific Alfa Aesar H56310.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-48-8 | C22H14 | 278.35 g/mol

Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

Solubility
Soluble in organic solvents.

Notes
Air and light sensitive. Store at cool temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-48-8
Molecular Formula C22H14
Molecular Weight (g/mol) 278.35
MDL Number MFCD00003710
InChI Key SLIUAWYAILUBJU-UHFFFAOYSA-N
Synonym benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
PubChem CID 8671
ChEBI CHEBI:33148
IUPAC Name pentacene
SMILES C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1

Specifications

Melting Point 372°C to 374°C (sublimation)
Quantity 1 g
Beilstein 1912418
Sensitivity Air and light sensitive
Merck Index 14,7107
Solubility Information Soluble in organic solvents.
Formula Weight 278.35
Percent Purity 97%
Chemical Name or Material Pentacene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.