missing translation for 'onlineSavingsMsg'
Learn More

n-Pentacontane, SPEX CertiPrep™

Product Code. 15216226
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
n-Pentacontane6596-40-31000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 6596-40-3
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1
Quantity 1 mL
CAS Max % 0.1
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C50H102
InChI Key PFLUOWJPZLHUEA-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
IUPAC Name pentacontane
Molecular Weight (g/mol) 703.366
PubChem CID 81067
Solvent or Matrix CS2:Pentane:MeCl
Chemical Name or Material n-Pentacontane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Pentacontane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : carbon disulphide, pentane, dichloromethane

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'