missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol, 98%, Thermo Scientific Chemicals
19.90 € - 116.00 €
Chemical Identifiers
CAS | 115-77-5 |
---|---|
Molecular Formula | C5H12O4 |
Molecular Weight (g/mol) | 136.15 |
InChI Key | WXZMFSXDPGVJKK-UHFFFAOYSA-N |
Synonym | pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek |
PubChem CID | 8285 |
IUPAC Name | 2,2-bis(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10234940
|
Thermo Scientific Acros
129872500 |
250 g | Plastic bottle |
19.90 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051300
|
Thermo Scientific Acros
129870010 |
1 kg | Plastic bottle |
35.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984631
|
Thermo Scientific Acros
129870050 |
5 kg | Plastic Drum |
116.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
115-77-5 | |
136.15 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
2,2-bis(hydroxymethyl)propane-1,3-diol |
C5H12O4 | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
8285 | |
C(C(CO)(CO)CO)O |
Specifications
115-77-5 | |
White | |
240°C | |
97.5% min. (GC) | |
C(CH2OH)4 | |
15,7221 | |
Solubility in water: 1 g/18 mL water (15°C). Other solubilities: slightly soluble in alcohol and formamide, insoluble in benzene, carbon tetrachloride, ether,, acetone and petroleum ether | |
C(C(CO)(CO)CO)O | |
136.15 | |
136.15 | |
Crystals |
257.0°C to 264.0°C | |
276.0°C (30.0 mmHg) | |
Authentic | |
C5H12O4 | |
01,528 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
2,2-bis(hydroxymethyl)propane-1,3-diol | |
8285 | |
98% | |
Pentaerythritol |
Safety and Handling
missing translation for 'einecsNumber' : 204-104-9
RUO – Research Use Only