Learn More
Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent.
Brand: Thermo Scientific™ TS-58220
313.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent. Reaction times are fast (∼20 minutes). Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids. PFBBr has been applied in analyzing trace organics in asphalts and as a fingerprinting technique for identifying asphalt pollutants found in surface waters.
- Fast reaction times for extraction alkylation technique: ∽20 minutes
- Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids
- Analysis of trace organics in asphalt
Recommended for:
Analysis of trace organics in asphalt
Specifications
GC | |
Room Temperature | |
98 to 100% |
Corrosive | |
alpha-Bromo-2,3,4,5,6-pentafluorotoluene | |
5 g |
Safety and Handling
- Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : alpha-bromo-2,3,4,5,6-pentafluorotoluene
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.