missing translation for 'onlineSavingsMsg'
Learn More
Pentafluorophenol, 99+%, Thermo Scientific Chemicals Product Code.: 10113290

Pentafluorophenol, 99+%, Thermo Scientific Chemicals

Product Code. 10113290
5 g, Glass bottle
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10113290

Brand: Thermo Scientific Acros 147130050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-61-9
Molecular Formula C6HF5O
Molecular Weight (g/mol) 184.07
MDL Number MFCD00002156
InChI Key XBNGYFFABRKICK-UHFFFAOYSA-N
Synonym pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp
PubChem CID 13041
IUPAC Name 2,3,4,5,6-pentafluorophenol
SMILES OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 34°C to 36°C
Color White
Boiling Point 143°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6F5OH
Quantity 5 g
Merck Index 15, 7223
Solubility Information Solubility in water: soluble
Formula Weight 184.07
Percent Purity 99+%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Pentafluorophenol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentafluorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pentafluorophenol, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.