missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 388.29 |
Percent Purity | 97% |
Chemical Name or Material | Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate |
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'