missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals

Product Code. 15790869
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3AgF5O2
Molecular Weight (g/mol) 270.89
MDL Number MFCD00004170
InChI Key XAYJXAUUXJTOSI-UHFFFAOYSA-M
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
SMILES [Ag+].[O-]C(=O)C(F)(F)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.5760g/mL
Boiling Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula CF3CF2CO2H
Refractive Index 1.2830 to 1.286
Quantity 250 g
Specific Gravity 1.576
IUPAC Name silver(1+) pentafluoropropanoate
Formula Weight 164.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'