Learn More
Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals
Description
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.
Solubility
Reacts with water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications
Specifications
Density | 1.589 |
Boiling Point | 73°C to 74°C |
Quantity | 25 g |
UN Number | UN3265 |
Beilstein | 1806446 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 310.04 |
Percent Purity | 98% |
Chemical Name or Material | Pentafluoropropionic anhydride |
Safety and Handling
- Pentafluoropropionic anhydride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Pentafluoropropionicanhydride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.