missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

67.54 € - 774.86 €

Chemical Identifiers

CAS 356-42-3
Molecular Formula C6F10O3
Molecular Weight (g/mol) 310.04
InChI Key XETRHNFRKCNWAJ-UHFFFAOYSA-N
Synonym pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
PubChem CID 67742
IUPAC Name 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
SMILES C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10521892
View Documents Promotion Details
Thermo Scientific Acros
416930050
5 g Glass bottle
67.54 €
5g
Please to purchase this item. Need a web account? Register with us today!
10529292
View Documents Promotion Details
Thermo Scientific Acros
416930250
25 g Glass bottle
227.13 €
25g
Please to purchase this item. Need a web account? Register with us today!
11964941
View Documents Promotion Details
Thermo Scientific Acros
416931000
100 g Glass bottle
774.86 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
Specifications

Chemical Identifiers

356-42-3
310.04
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
C6F10O3
XETRHNFRKCNWAJ-UHFFFAOYSA-N
67742
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F

Specifications

356-42-3
1.5700g/mL
Authentic
C6F10O3
1.3
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
310.04
310.04
Liquid
Colorless
69.0°C
97.5% min. (GC)
(CF3CF2CO)2O
1.57
XETRHNFRKCNWAJ-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
67742
98%
Pentafluoropropionic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-604-2

Product Identifier
  • Pentafluoropropionic anhydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only