missing translation for 'onlineSavingsMsg'
Learn More
Pentyl isocyanate, 98%, Thermo Scientific Chemicals Product Code.: 10399260

Pentyl isocyanate, 98%, Thermo Scientific Chemicals

Product Code. 10399260
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10399260

Brand: Thermo Scientific Acros 314320050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3954-13-0
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00144846
InChI Key VRVUKQWNRPNACD-UHFFFAOYSA-N
Synonym pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhgShow MoreShow Less
PubChem CID 206036
IUPAC Name 1-isocyanatopentane
SMILES CCCCCN=C=O

Specifications

Color Colorless
Density 0.8700g/mL
Boiling Point 136.0°C to 137.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4NCO
Refractive Index 1.4130 to 1.4150
Quantity 5 g
Specific Gravity 0.87
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pentyl isocyanate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number098143I Certificate TypeCertificate of Analysis Date04/04/2025 Catalog Number
Lot Number098143I Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number
Lot Number098143I Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number
Lot Number098143I Certificate TypeCertificate of Analysis Date14/02/2025 Catalog Number
Lot NumberA0375647 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number314320010,314320050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pentyl isocyanate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.