Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
394.91 € - 546.11 €
Specifications
For Use With | PepMap™ Neo Trap Cartridge (part number 174500) |
---|---|
Max. Pressure | 22,000 psi (1500 bar) |
Temperature | 60°C |
Product Line | PepMap Neo |
For Use With (Application) | Trap and Elute, Peptide Mapping |
Product Code | Brand | Description | Length (Metric) | For Use With | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Length (Metric) | For Use With | Price | Quantity & Availability | |||
17427583
|
Thermo Scientific™
174501 |
PEEK Tubing with nanoViper™ Fittings | 100 mm | PepMap™ Neo Trap Cartridge (part number 174500) |
394.91 €
Pack of 2 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17437583
|
Thermo Scientific™
174502 |
Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings | 5 mm | PepMap™ Neo Trap Cartridge (part number 174500) |
546.11 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
Description
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
Specifications
PepMap™ Neo Trap Cartridge (part number 174500) | |
60°C | |
Trap and Elute, Peptide Mapping |
22,000 psi (1500 bar) | |
PepMap Neo |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.