missing translation for 'onlineSavingsMsg'
Learn More

Pepsin-HCl Electrode Cleaning Solution, Reagecon™
BIOPHARMA

Ideal for removing protein contamination from the permeable diaphragm of pH electrodes. Pepsin-HCl Electrode Cleaning Solution, Reagecon™ is a solution of at least 1% pepsin A in 0.1M hydrochloric acid. Electrode elements should be soaked for approximately one hour.

Brand:  Reagecon 90508


Product Code. 11726378

  • 46.45 € / 250mL
Estimated Shipment: 11-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Pepsin A is an enzyme that breaks down proteins into smaller peptides (a protease). It is produced naturally in the human digestive system and is produced commercially for use in laboratories and in the food industry.
Specifications

Specifications

100°C
Pepsin A in Hydrochloric Acid
Pepsin
0°C
250 mL
Colorless
pH Electrode
Electrode cleaning, removal of protein contamination
Odorless
Fully miscible in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SOLUTION DE NETTOYAGE

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Pepsin A

SDS
Documents

Documents

Certificates
Special Offers

Special Offers