Learn More
Novus Biologicals™ Pepsin Solution
Brand:  Novus Biologicals™ NB900-66724
Product Code. 18298053
  • 464.00 € / 50mL
Explore more special offers
Specifications
4°C. Do not freeze.
50 mL
Preparation of Working Solutions 1. Solution Pepsin is Ready to Use and Does Not Require Any Preparation. Protocol Recommendations 1. Deparaffinize Tissue Sections and Wash Slides with Wash Buffer. 2. Remove Excess Buffer From Slide Without Letting The Tissue Dry. 3. Depending on The Size of The Tissue Section Add 1-2 Drops of Pepsin Solution. 4. Incubate Slide for 5-10 Minutes (at 37 C. 5. Wash Slide with Wash Buffer and Proceed with Immunostaining.)
Pepsin Solution