Learn More
Fisherbrand™ Ion Selective Electrode
Ion selective perchlorate electrode with a reproducibility greater than 2%.
Brand:  Fisherbrand™ PER1522-S7
Product Code. 11719718
  • 580.00 € / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ion selective perchlorate electrode
  • Reproducibility better than 2%
  • Interchangeable membrane
  • S7 compliant per-head connection
  • All pH / mV meters or ionometers
Specifications
Specifications
Ion Selective Electrode
2.5 to 11
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products