missing translation for 'onlineSavingsMsg'
Learn More
Perchloric acid, 70%, 99.9985% (metals basis), Thermo Scientific Chemicals Product Code.: 15420997

Perchloric acid, 70%, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 15420997
50 mL
Click to view available options
Quantity:
5 mL
50 mL
250 mL
This item is not returnable. View return policy

Product Code. 15420997

Brand: Thermo Scientific Alfa Aesar 010983.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perchloric acid: 70%

Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis. It finds application in the computer industry for etching liquid crystal displays in critical electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Appearance Colorless liquid
Color White
Melting Point -112°C
Boiling Point 203°C
Density 1.67 g/cm3 at 20°C
Odor Pungent
Vapor Pressure 9 hPa (7mm Hg)
Flash Point 113°C (235°F)
Assay Percent Range 99.9985% (metals basis)
Quantity 50 mL
Physical Form Liquid
UN Number 1873
Solubility Information Miscible with water.
Formula Weight 100.46
Concentration or Composition (by Analyte or Components) Total Metal Impurities: 0.0015% max.Concentration (weight/weight): 70% min
Chemical Name or Material Perchloric Acid 70%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberTN752482 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number010983.AA,010983.AD,010983.AK
Lot NumberGZ730393 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number010983.AA,010983.AD,010983.AK
Lot Number10244616 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog Number010983.AA,010983.AD,010983.AK
Lot Number10242089 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number010983.AA,010983.AD,010983.AK
Lot NumberI13P07A Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010983.AA,010983.AD,010983.AK
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Perchloric acid, 70%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Perchloric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perchloric acid, 70%, 99.9985% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.