missing translation for 'onlineSavingsMsg'
Learn More
Perchloric acid, ACS, 48-50%, Thermo Scientific Chemicals Product Code.: 15461997

Perchloric acid, ACS, 48-50%, Thermo Scientific Chemicals

Product Code. 15461997
2.5 L
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
This item is not returnable. View return policy

Product Code. 15461997

Brand: Thermo Scientific Alfa Aesar 044464.K4

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm

Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point °C to 112°C
Density 1.41
Vapor Pressure 29mm Hg at 20°C
Assay Percent Range 48-50%
Quantity 2.5 L
Physical Form Liquid
UN Number UN1802
Solubility Information Miscible with water.
Formula Weight 100.46
Grade ACS
Chemical Name or Material Perchloric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27L043 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number044464.AE,044464.K4,044464.AP
Lot NumberM27L027 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number044464.AE,044464.K4,044464.AP
Lot NumberY04K046 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number044464.K4,044464.AP
Lot NumberT11K038 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number044464.K4,044464.AP
Lot NumberP20F026 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number044464.K4,044464.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perchloric acid, ACS, 48-50%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.