missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Description
ClHO4, CAS Number-7601-90-3, unii-v561v90bg2, ueberchlorsaeure, perchlorsaeure, perchloric acid, hclo4, hyperchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchloric acid, 1L, CHEBI:29221, Colorless, VLTRZXGMWDSKGL-UHFFFAOYSA-N, 11325, 100.454

Specifications
Specifications
pH | 0.1 |
Color | Colorless |
Packaging | HDPE plastic bottle |
Quantity | 2.5 L |
Physical Form | Liquid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526159 | Certificate of Analysis | 11/02/2025 | J/5820/PB17,J/5820/24,J/5820/25,J/5820/PB15 |
Safety and Handling


Product Identifier
- Perchloric acid, 0.1 M solution in acetic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction