missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide solution, PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur., Honeywell Fluka™

Product Code. 15613490
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur.

Honeywell ACS solvents for general laboratory use are:

  • a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested tmeet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

CAS Min % 68.9000%, 30.0000%
CAS Max % 69.9000%, 31.0000%
Linear Formula H2O2
UN Number UN2014
Beilstein 3587191
Density 1.11 g/cm3
Formula Weight 34.01
Percent Purity >30%
Grade puriss. p.a.
Packaging Poly Bottle
Quantity 6 x 1 L
Chemical Name or Material Hydrogen peroxide solution, PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • PERDROGEN 30% H2O2 (w/w)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.