missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals

Product Code. 11487044
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-76-5
Molecular Formula C9F18
Molecular Weight (g/mol) 450.07
MDL Number MFCD00066614
InChI Key MGOFOLPXKULBGG-UHFFFAOYSA-N
Synonym perfluoro-1,3,5-trimethylcyclohexane, 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl cyclohexane, acmc-1ajc4, nonafluoro-1,3,5-tris-trifluoromethyl-cyclohexane, 1,2,2,3,4,4,5,6,6-nonafluoro-1,3,5-tris trifluoromethyl cyclohexane, cyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl, 1,1,2,3,3,4,5,5,6-nonakis fluoranyl-2,4,6-tris trifluoromethyl cyclohexane
PubChem CID 2776392
IUPAC Name 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -68°C
Density 1.888
Boiling Point 125°C to 128°C
Refractive Index 1.297
Quantity 10 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 450.07
Grade Technical
Chemical Name or Material Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'