missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(1,3-dimethylcyclohexane), tech. 90%, Thermo Scientific Chemicals

Product Code. 15432297
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15432297

Brand: Thermo Scientific Alfa Aesar 045477.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 335-27-3
Molecular Formula C8F16
Molecular Weight (g/mol) 400.062
MDL Number MFCD00001469
InChI Key LOQGSOTUHASIHI-UHFFFAOYSA-N
Synonym perfluoro-1,3-dimethylcyclohexane, perfluoro 1,3-dimethylcyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl cyclohexane, decafluoro-1,3-bis trifluoromethyl cyclohexane, cyclohexane, perfluoro-1,3-dimethyl, cyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl, cyclohexane, decafluoro-1,3-bis trifluoromethyl, hexadecafluoro 1,3-dimethylcyclohexane, flutec pp3, acmc-1cjoy
PubChem CID 78975
IUPAC Name 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifikationer

Melting Point -55°C
Density 1.828 g/mL
Boiling Point 101°C to 102°C
Odor Odorless
Quantity 250 g
Beilstein 2064147
Formula Weight 400.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Perfluoro (1,3-dimethylcyclohexane)
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.