missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-butanesulfonyl fluoride, 92%, Thermo Scientific Chemicals

Product Code. 10349410
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-72-4
Molecular Formula C4F10O2S
Molecular Weight (g/mol) 302.09
MDL Number MFCD00007422
InChI Key LUYQYZLEHLTPBH-UHFFFAOYSA-N
Synonym nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
PubChem CID 67814
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
SMILES C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F

Specifications

Color Colorless to Yellow
Density 1.6800g/mL
Boiling Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)3SO2F
Refractive Index 1.3
Quantity 25 g
Specific Gravity 1.68
Formula Weight 302.09
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Perfluoro-1-butanesulfonyl fluoride, 92%
Show More Show Less
compliance-icons
Product Identifier
  • Perfluoro-1-butanesulfonyl fluoride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.