missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-butanesulfonyl fluoride, 92%, Thermo Scientific Chemicals

Product Code. 10668444
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-72-4
Molecular Formula C4F10O2S
Molecular Weight (g/mol) 302.09
MDL Number MFCD00007422
InChI Key LUYQYZLEHLTPBH-UHFFFAOYSA-N
Synonym nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
PubChem CID 67814
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
SMILES C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F

Specifications

Color Colorless to Yellow
Density 1.6800g/mL
Boiling Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)3SO2F
Refractive Index 1.3
Quantity 5 g
Specific Gravity 1.68
Formula Weight 302.09
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Perfluoro-1-butanesulfonyl fluoride, 92%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'