missing translation for 'onlineSavingsMsg'
Learn More
Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals Product Code.: 11413837

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Product Code. 11413837
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11413837

Brand: Thermo Scientific Alfa Aesar A18288.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.081
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Especificaciones

Melting Point -10°C to -7°C
Density 1.94
Boiling Point 141°C to 142°C
Refractive Index 1.3145
Quantity 25 g
Beilstein 2067113
Merck Index 14,4183
Solubility Information Insoluble in water.
Formula Weight 462.1
Percent Purity 95%
Chemical Name or Material Perfluoro(decahydronaphthalene), cis + trans
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242657 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberA18288.22,A18288.14,A18288.36
Lot Number10167929 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18288.22,A18288.14,A18288.36
Lot Number10166111 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18288.22,A18288.14,A18288.36
Lot Number10223010 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18288.22,A18288.14,A18288.36
Lot Number10054333 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18288.22,A18288.14,A18288.36

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado