Learn More
Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals
Description
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | -10°C to -7°C |
Density | 1.94 |
Boiling Point | 141°C to 142°C |
Refractive Index | 1.3145 |
Quantity | 100 g |
Beilstein | 2067113 |
Merck Index | 14,4183 |
Solubility Information | Insoluble in water. |
Formula Weight | 462.1 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.