Learn More
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Shop All Thermo Scientific ProductsBeskrivelse
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI). Compared to acid chlorides and anhydrides, the reactions of fluorinated acylimidazoles are smooth and quantitative, and produce no acid byproducts. The principal byproduct is imidazole, which is relatively inert. The volatile and stable fluoroacyl derivatives are ideal for FID and electron-capture detection (ECD) techniques.
Used in bifunctional derivatization schemes and in exchange reactions in which TMS derivatives are converted to HFB derivatives.
Hydroxyl groups of catecholamines are derivatized with TMSI, followed by conversion of the amines to acylamides with HFBI.
Tryptamine and metabolites present in spinal fluid have been analyzed by ECD using HFBI.
Tekniske data
Tekniske data
| For Use With | FID, ECD |
| Hazards | Corrosive, Flammable |
| Storage | 2°C to 8°C |
| Quantity | 10 x 1 mL Ampules |
| Standard Components | TFAI |
| Standard Concentration | 98 to 100% |
| Item Description | TFAI; 10 x 1 mL ampules |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.