missing translation for 'onlineSavingsMsg'
Learn More
Perfluorodecalin, 90%, mixture of cis and trans, Thermo Scientific Chemicals Product Code.: 10626724

Perfluorodecalin, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 10626724
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10626724

Brand: Thermo Scientific Acros 130041000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.08
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latinShow MoreShow Less
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Specifications

Melting Point -10.0°C
Color Colorless
Density 1.9000g/mL
Boiling Point 142.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.3135 to 1.3155
Quantity 100 g
Merck Index 15,4215
Specific Gravity 1.9
Solubility Information Solubility in water: insoluble
Formula Weight 462.08
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Perfluorodecalin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463138 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number130040250,130045000,130041000
Lot NumberA0455126 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number130040250,130045000,130041000
Lot NumberA0403435 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number130040250,130045000,130041000
Lot NumberA0400411 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number130040250,130045000,130041000
Lot NumberA0393483 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number130040250,130045000,130041000
compliance-icons
Product Identifier
  • Perfluorodecalin, mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perfluorodecalin, 90%, mixture of cis and trans, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.