missing translation for 'onlineSavingsMsg'
Learn More

Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11465966
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-76-2
Molecular Formula C10HF19O2
Molecular Weight (g/mol) 514.09
MDL Number MFCD00004175
InChI Key PCIUEQPBYFRTEM-UHFFFAOYSA-N
Synonym perfluorodecanoic acid, nonadecafluorodecanoic acid, ndfda, pfda, perfluoro-n-decanoic acid, nonadecafluoro-n-decanoic acid, decanoic acid, nonadecafluoro, ccris 4417, decanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro, perfluorocapric acid
PubChem CID 9555
ChEBI CHEBI:35546
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 77°C to 82°C
Boiling Point 218°C
Linear Formula CF3(CF2)8CO2H
Quantity 25 g
UN Number UN2923
Beilstein 1810811
Formula Weight 514.09
Percent Purity 97%
Chemical Name or Material Perfluorodecanoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'