missing translation for 'onlineSavingsMsg'
Learn More

Perfluoroheptanoic acid, 96%, Thermo Scientific Chemicals

Product Code. 10043380
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 375-85-9
Melting Point 30.0°C
Color Colorless to Yellow
Boiling Point 175.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C7HF13O2
Linear Formula CF3(CF2)5COOH
Refractive Index 1.3045 to 1.3075
MDL Number MFCD00039604
Quantity 10 g
Beilstein 04, 965
Synonym perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
InChI Key ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
SMILES C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
Molecular Weight (g/mol) 364.05
PubChem CID 67818
ChEBI CHEBI:35547
Formula Weight 364.05
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material Perfluoroheptanoic acid, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perfluoroheptanoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'