missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanes, 98+%, Thermo Scientific Chemicals

Product Code. 11317148
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluorohexanes is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. It plays a vital role as a reaction medium, especially for photooxidation reactions. Further, it finds application as an ultrasound contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-42-0
Molecular Formula C6F14
Molecular Weight (g/mol) 338.044
MDL Number MFCD00000437
InChI Key ZJIJAJXFLBMLCK-UHFFFAOYSA-N
Synonym perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
PubChem CID 9639
ChEBI CHEBI:39427
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -4°C
Color Colorless
Density 1.699 g/mL
Boiling Point 58°C to 60°C
Flash Point None
Linear Formula CF3(CF2)4CF3
Refractive Index 1.251
Quantity 100 g
Beilstein 1802113
Solubility Information Immiscible with water.
Formula Weight 338.04
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Perfluorohexanes
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.