missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanoyl chloride, 97%, Thermo Scientific™

Product Code. 11377326
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Perfluorooctanoyl chloride is derivatizated and used in the determination of Amphetamine and Methamphetamine in Blood. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents and moisture. Store under dry inert gas. Incompatible with strong oxidizing agents, strong bases, alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-64-8
Molecular Formula C8ClF15O
Molecular Weight (g/mol) 432.51
MDL Number MFCD00013654
InChI Key AQQBRCXWZZAFOK-UHFFFAOYSA-N
Synonym perfluorooctanoyl chloride, pentadecafluorooctanoyl chloride, pentadecafluorooctyl chloride, octanoyl chloride, pentadecafluoro, perfluorooctanoylchloride, perfluorooctanyl chloride, perflurooctanoyl chloride, acmc-1cmee, perfluorocaprylic chloride, n-perfluorooctanoyl chloride
PubChem CID 78978
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O

Specifications

Density 1.744
Boiling Point 130°C to 132°C
Refractive Index 1.3045
Quantity 25 g
UN Number UN3265
Beilstein 1809677
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name pentadecafluorooctanoyl chloride
Formula Weight 432.52
Percent Purity 97%
Chemical Name or Material Perfluorooctanoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.