Learn More
Heidolph Instruments Perforated Platform 2000
For use with Unimax 2010, Promax 2020 and Polymax 2040
Brand:  Heidolph Instruments 549-59000-00
Product Code. 10613211
  • 205.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Universal perforated platform allows for individual arrangement of vessels
Specifications
Specifications
0 to 1100 rpm
Unimax 2010, Promax 2020 and Polymax 2040