missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 126.11 |
Percent Purity | 97% |
Chemical Name or Material | Perhydrocyclobuta[c]furan-1, 3-dione |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction