missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ PeridoxRTU

PeridoxRTU is a blend of hydrogen peroxide and peracetic acid.
259.00 € - 271.00 €
Specifications
Product Type | Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner |
---|---|
Form | Liquid |
pH | Not Applicable |
Scent | Unscented |
Sterility | Non-sterile |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17241502
|
Contec™
FBC0902PX |
0.9 L |
259.00 €
Pack of 6 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
17251502
![]() |
Contec™
FBC3702PX |
3.78 L |
271.00 €
Pack of 4 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
Description
- Contec PeridoxRTU is a BPR and EPA authorised sporicide achieving greater than a log 3 reduction against bacterial and fungal spores in under 3 min
- A blend of hydrogen peroxide and peracetic acid, Contec PeridoxRTU is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts and spores
- PeridoxRTU's proprietary formulation contains surfactants for cleaning efficiency along with corrosion inhibitors for surface protection
- PeridoxRTU can be used to clean and disinfect surfaces without damage or loss of functionality
- Contec PeridoxRTU is filtered and filled in a controlled environment
- The sterile product is provided validated sterile to a Sterility Assurance Level of 10-6
- Provided double or single bagged, the product is designed for ease of entry into pharmaceutical cleanrooms
- Available as either a 0.9 L bottle with a push/pull cap for small areas or in 3.78 L capped containers for mop head saturation or when larger volumes are required
- Patented hyperactive chemistry makes it fast acting so save time spent of biodecontamination
- Validated sterile version available makes it suitable for Grade A and B cleanrooms
- Excellent material compatibility and will not damage common cleanroom materials
- Double/single bagged helps facilitate transfer disinfection into cleanroom
- Push/pull cap on 0.9 L eliminates droplets reducing the risk of inhalation and can be easily applied to a wipe or cleaning too
Specifications
Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner | |
Not Applicable | |
Non-sterile | |
Not Autoclavable | |
Hydrogen Peroxide and Peracetic Acid | |
Limited-use | |
No |
Liquid | |
Unscented | |
Clear | |
Clear | |
RTU | |
Non-filtered |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- PeridoxRTU
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, hydrogen peroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title