missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10592981
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White to Yellow
Melting Point 122.0°C
Identification Positive for periodate
Packaging Glass bottle
Assay Percent Range 99% min. (Iodometry)
Quantity 100 g
Linear Formula H5IO6
Physical Form Crystals or Crystalline Powder
Fieser 01,815; 02,313; 03,220; 04,374; 05,508; 10,304; 13,238; 16,272
Merck Index 15, 7286
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol,slightly soluble in ether
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Formula Weight 227.94
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Periodic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Periodic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'