missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, 99%, Thermo Scientific Chemicals

Product Code. 11434387
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

Solubility
Soluble in water and alcohols.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, strong bases and dimethyl sulfoxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
SMILES O[I](O)(O)(O)(O)=O

Specifications

pH 1.2
Melting Point 126°C to 130°C
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Quantity 1000 g
UN Number UN3085
Merck Index 14,7171
Solubility Information Soluble in water and alcohols.
Formula Weight 227.94
Percent Purity 99%
Chemical Name or Material Periodic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'