missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Certified AR for Analysis, Fisher Chemical

Product Code. 10467260
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 122°C
CAS 10450-60-9
Color White
pH 1
Physical Form Solid
Quantity 100 g
Molecular Formula H5IO6
MDL Number MFCD00011344
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES O[I](O)(O)(O)(O)=O
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Molecular Weight (g/mol) 227.94
PubChem CID 25289
ChEBI CHEBI:29150
Formula Weight 227.94g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Periodic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.