missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow 120 Series Drive with Pumphead

120 series pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.

3795.00 € - 11175.00 €

Specifications

Display Type LED
View More Specs

Products 6
Product Code Brand Model Flow Rate Motor Speed Price Quantity & Availability  
Product Code Brand Model Flow Rate Motor Speed Price Quantity & Availability  
12555316
View Documents
Watson-Marlow
010.6141.R00
120U/R 0.003 to 54 mL/min. 0.1 to 32 rpm
3795.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12466260
View Documents
Watson-Marlow
010.6131.DA0
120S/DV 0.001 to 170 mL/min. at 30 psi 1 to 200 rpm
6405.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12426260
View Documents
Watson-Marlow
010.6131.010
120S/D1 0.001 to 120 mL/min. 0.01 to 200 rpm
6425.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12446260
View Documents
Watson-Marlow
010.6131.M30
120S/DM3 0.0001 to 36 mL/min. 0.01 to 100 rpm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12436260
View Documents
Watson-Marlow
010.6131.M20
120S/DM2 0.0001 to 36 mL/min. 0.01 to 100 rpm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12476260
View Documents
Watson-Marlow
010.6141.DA0
120U/DV 0.002 to 190 mL/min. 0.1 to 200 rpm
9710.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The 120 series is the perfect pump for maximising yield in fermentation and cell culture applications. Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimise your process. The 120 series pump is perfectly suited to single-use systems. Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate. With an exceptional 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials. A keypad lock secures your pumping parameters for consistent and repeatable performance.
  • Pumphead options available as standard (others available on request):

  • 120R single channel, two roller pumphead for continuous tubing
Specifications

Specifications

LED
Videos
SDS
Documents

Documents

Product Certifications