Learn More
Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Manufactured to the utmost standard. Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads are autoclavable.
101.00 € - 258.00 €
Specifications
Overall Length (Metric) | 40 cm |
---|---|
For Use With | MF1 and MF4 medium flow pump head |
For Use With (Application) | Medium flow tubing has color coded retaining stops for easy recognition of the tubing's internal diameter |
Sterility | Sterile |
Product Type | Peristaltic Pump Tubing |
Product Code | Brand | Flow Rate | Gas Permeability | Color | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Gas Permeability | Color | Material | Price | Quantity & Availability | |||
10205692
|
Gilson™
F117934 |
1.42mL/min. | Low | Orange/Yellow | PVC |
106.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||
10021972
|
Gilson™
F117956 |
3mL/min. | Low | Black | PVC |
106.00 €
3m |
In Stock
Log in to see stock. |
|||
10153192
|
Gilson™
F117960 |
9mL/min. | Low | Gray | PVC |
101.00 €
3m |
In Stock
Log in to see stock. |
|||
10133002
|
Gilson™
F117943 |
16mL/min. | Low | Blue | PVC |
106.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||
10694153
|
Gilson™
F117942 |
14mL/min. | Low | Yellow/Blue | PVC |
106.00 €
Pack of 12 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
|||
10285922
|
Gilson™
F117963 |
16mL/min. | Low | Blue | PVC |
101.00 €
3m |
In Stock
Log in to see stock. |
|||
10357422
|
Gilson™
F1825112 |
1mL/min. | High | White | Silicone |
126.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||
10377752
|
Gilson™
F117945 |
25mL/min. | Low | Purple | PVC |
101.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||
10082002
|
Gilson™
F117966 |
29mL/min. | Low | Purple/Black | PVC |
101.00 €
3m |
In Stock
Log in to see stock. |
|||
10387752
|
Gilson™
F117946 |
29mL/min. | Low | Violet/Black | PVC |
106.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||
10042052
|
Gilson™
F1825114 |
2.8mL/min. | High | Violet/White | Silicone |
126.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||
10226122
|
Gilson™
F1825113 |
2mL/min. | High | Pink/Pink | Silicone |
126.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||
10760095
|
Gilson™
F117949 |
47mL/min. | Low | Black/White | PVC |
106.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||
10367612
|
Gilson™
F117747 |
55mL/min. | Impermeable to most gases | Black | Iso-Versinic |
232.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||
10695883
|
Gilson™
F117749 |
70mL/min. | Impermeable to most gases | Black | Iso-Versinic |
258.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||
Description
As part of our MINIPULS™ 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Choice of Materials
- Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
- PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications
40 cm | |
Medium flow tubing has color coded retaining stops for easy recognition of the tubing's internal diameter | |
Peristaltic Pump Tubing |
MF1 and MF4 medium flow pump head | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads