missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ PerkinElmer Split / Splitless FocusLiners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 092059

 View more versions of this product

Product Code. 10309142

  • 211.00 € / Pack of 5
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimadzu instruments
Specifications

Specifications

5/Pk.
GC Inlet Liner
5 mm
95 mm
3.4 mm
Shimadzu 2010 GC Instruments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers